Assay Details
Target Gene Details
Entrez Gene ID: | 55904 |
Gene Name: | lysine methyltransferase 2E |
Gene Aliases: |
HDCMC04P, MLL5, NKp44L |
Location: |
Chr.7:105014190-105114085 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
KMT2E | NM_018682.3 | NP_061152.3 | ||
NM_182931.2 | NP_891847.1 | |||
XM_005250493.1 | XP_005250550.1 | |||
XM_011516400.1 | XP_011514702.1 | |||
XM_017012435.1 | XP_016867924.1 | |||
AF519459.1 | AAM74947.1 | |||
AK000940.1 | ||||
AY147037.1 | AAN17675.1 | |||
BC001296.2 | AAH01296.1 | |||
BC040004.1 | AAH40004.1 | |||
BC053906.1 | AAH53906.1 | |||
BC062583.1 | AAH62583.1 | |||
BC142987.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv608068 | Chr.7:104981955 - 105117036 on Build GRCh38 | Loss | KMT2E-AS1 SRPK2 LINC01004 KMT2E LOC101927902 |
nsv436902 | Chr.7:104802439 - 105205347 on Build GRCh38 | Insertion | LHFPL3-AS2 KMT2E-AS1 LHFPL3-AS1 SRPK2 LINC01004 KMT2E LOC101927902 LHFPL3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |