Assay Details
Target Gene Details
Entrez Gene ID: | 83943 |
Gene Name: | inner mitochondrial membrane peptidase subunit 2 |
Gene Aliases: |
IMMP2L-IT1, IMP2, IMP2-LIKE |
Location: |
Chr.7:110662645-111562531 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IMMP2L | NM_001244606.1 | NP_001231535.1 | ||
NM_032549.3 | NP_115938.1 | |||
XM_005250630.3 | XP_005250687.1 | |||
XM_011516605.2 | XP_011514907.1 | |||
XM_011516609.2 | XP_011514911.1 | |||
XM_011516613.2 | XP_011514915.1 | |||
XM_017012705.1 | XP_016868194.1 | |||
AF359563.1 | AAK52905.1 | |||
AK311968.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3614527 | Chr.7:110564413 - 110700754 on Build GRCh38 | Gain | IMMP2L |
nsv608147 | Chr.7:110651990 - 110977789 on Build GRCh38 | Loss | IMMP2L |
esv3576414 | Chr.7:110485735 - 110864936 on Build GRCh38 | Gain | IMMP2L |
nsv608080 | Chr.7:109461248 - 111666297 on Build GRCh38 | Gain | LRRN3 EIF3IP1 IMMP2L |
dgv11570n54 | Chr.7:110662639 - 110702956 on Build GRCh38 | Gain | IMMP2L |
nsv1016827 | Chr.7:110661290 - 110706914 on Build GRCh38 | Gain | IMMP2L |
dgv6564n100 | Chr.7:110589001 - 110749639 on Build GRCh38 | Loss | IMMP2L |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |