Assay Details
Target Gene Details
Entrez Gene ID: | 6677 |
Gene Name: | sperm adhesion molecule 1 |
Gene Aliases: |
HEL-S-96n, HYA1, HYAL1, HYAL3, HYAL5, PH-20, PH20, SPAG15 |
Location: |
Chr.7:123925232-123971410 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SPAM1 | NM_001174044.1 | NP_001167515.1 | ||
NM_001174045.1 | NP_001167516.1 | |||
NM_001174046.1 | NP_001167517.1 | |||
NM_003117.4 | NP_003108.2 | |||
NM_153189.2 | NP_694859.1 | |||
AK292229.1 | ||||
AY920278.1 | AAX13985.1 | |||
AY920279.1 | AAX13986.1 | |||
AY920280.1 | AAX13987.1 | |||
AY920281.1 | AAX13988.1 | |||
BC026163.1 | AAH26163.1 | |||
BI831525.1 | ||||
DB023094.1 | ||||
EU794686.1 | ||||
S67798.1 | AAC60607.2 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)