Assay Details
Target Gene Details
Entrez Gene ID: | 1644 |
Gene Name: | dopa decarboxylase |
Gene Aliases: |
AADC |
Location: |
Chr.7:50458436-50565460 on Build GRCh38 |
Assay Gene Location: | Within Intron 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDC | NM_000790.3 | NP_000781.1 | ||
NM_001082971.1 | NP_001076440.1 | |||
NM_001242886.1 | NP_001229815.1 | |||
NM_001242887.1 | NP_001229816.1 | |||
NM_001242888.1 | NP_001229817.1 | |||
NM_001242889.1 | NP_001229818.1 | |||
XM_005271745.4 | XP_005271802.1 | |||
XM_011515161.2 | XP_011513463.2 | |||
AK298321.1 | ||||
AK298392.1 | ||||
BC000485.1 | AAH00485.1 | |||
BC008366.1 | AAH08366.1 | |||
BT007009.1 | AAP35655.1 | |||
CD013934.1 | ||||
CD013935.1 | ||||
CD013936.1 | ||||
CD013937.1 | ||||
CD013938.1 | ||||
CR456724.1 | CAG33005.1 | |||
KU178021.1 | ||||
KU178022.1 | ||||
M76180.1 | AAA58437.1 | |||
M88700.1 | AAA20894.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1016665 | Chr.7:50384854 - 51336827 on Build GRCh38 | Gain | COBL IKZF1 DDC-AS1 FIGNL1 GRB10 DDC |
nsv1026081 | Chr.7:50458202 - 50470486 on Build GRCh38 | Loss | DDC |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |