Assay Details
Target Gene Details
Entrez Gene ID: | 27297 |
Gene Name: | CGRP receptor component |
Gene Aliases: |
CGRP-RCP, CGRPRCP, RCP, RCP9 |
Location: |
Chr.7:66114818-66154568 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CRCP | NM_001040647.1 | NP_001035737.1 | ||
NM_001040648.1 | NP_001035738.1 | |||
NM_001142414.1 | NP_001135886.1 | |||
NM_014478.4 | NP_055293.1 | |||
NR_024548.1 | ||||
AF073792.1 | AAC25992.1 | |||
AK290553.1 | ||||
AK303737.1 | ||||
AK311829.1 | ||||
BC040107.1 | AAH40107.1 | |||
BC105808.1 | AAI05809.1 | |||
BI600330.1 | ||||
BX647867.1 | CAI46069.1 | |||
DA821665.1 | ||||
DA955978.1 | ||||
DB472426.1 | ||||
U51134.1 | AAD05036.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2761079 | Chr.7:65601260 - 66224746 on Build GRCh38 | Gain+Loss | SNORA22 LOC84214 LOC441242 TPST1 ASL INTS4P2 GUSB VKORC1L1 CCT6P1 CRCP |
esv2664145 | Chr.7:65814760 - 66395282 on Build GRCh38 | Deletion | LINC00174 LOC84214 TPST1 ASL GUSB VKORC1L1 CRCP |
nsv1021186 | Chr.7:66027614 - 66198451 on Build GRCh38 | Gain | ASL CRCP |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |