Assay Details
Target Gene Details
Entrez Gene ID: | 4852 |
Gene Name: | neuropeptide Y |
Gene Aliases: |
PYY4 |
Location: |
Chr.7:24284188-24291865 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
NPY | NM_000905.3 | NP_000896.1 | ||
AI198311.1 | ||||
AK312156.1 | ||||
BC029497.1 | AAH29497.1 | |||
K01911.1 | AAA59944.1 | |||
M15789.1 | AAA59946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2673027 | Chr.7:23679603 - 24432457 on Build GRCh38 | Deletion | LOC107986777 FAM221A NPY STK31 |
nsv830927 | Chr.7:24262646 - 24438857 on Build GRCh38 | Gain | LOC107986777 NPY |
nsv517053 | Chr.7:24224894 - 24386936 on Build GRCh38 | Gain+Loss | LOC107986777 NPY |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |