Assay Details
Target Gene Details
Entrez Gene ID: | 83605 |
Gene Name: | CCM2 scaffolding protein |
Gene Aliases: |
C7orf22, OSM, PP10187 |
Location: |
Chr.7:44999746-45076470 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCM2 | NM_001029835.2 | NP_001025006.1 | ||
NM_001167934.1 | NP_001161406.1 | |||
NM_001167935.1 | NP_001161407.1 | |||
NM_031443.3 | NP_113631.1 | |||
NR_030770.1 | ||||
XM_006715785.3 | XP_006715848.1 | |||
XM_006715786.3 | XP_006715849.1 | |||
XM_011515561.2 | XP_011513863.1 | |||
XM_011515562.1 | XP_011513864.1 | |||
XM_011515563.2 | XP_011513865.1 | |||
XM_011515564.2 | XP_011513866.1 | |||
XM_017012671.1 | XP_016868160.1 | |||
XM_017012672.1 | XP_016868161.1 | |||
XM_017012673.1 | XP_016868162.1 | |||
AF370392.1 | AAQ15228.1 | |||
AK098005.1 | ||||
AK298671.1 | ||||
AK303216.1 | ||||
AK303534.1 | ||||
BC004903.2 | AAH04903.1 | |||
BC008859.2 | AAH08859.1 | |||
BC016832.2 | AAH16832.1 | |||
BC063663.1 | AAH63663.1 | |||
DA420491.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |