Assay Details
Target Gene Details
Entrez Gene ID: | 223075 |
Gene Name: | coiled-coil domain containing 129 |
Gene Aliases: |
- |
Location: |
Chr.7:31514071-31658720 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC129 | NM_001257967.1 | NP_001244896.1 | ||
NR_047565.1 | ||||
XM_011515213.1 | XP_011513515.1 | |||
XM_017011871.1 | XP_016867360.1 | |||
XM_017011872.1 | XP_016867361.1 | |||
AK128026.1 | BAC87235.1 | |||
AK304387.1 | ||||
BC132719.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891094 | Chr.7:31500343 - 31591980 on Build GRCh38 | Gain | CCDC129 |
nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion | INMT-FAM188B LOC101928268 ZNRF2 LOC100130673 GHRHR LOC401320 PPP1R17 AVL9 DPY19L1P1 DKFZP586I1420 INMT ZNRF2P1 PDE1C NEUROD6 NOD1 MIR550A1 CRHR2 GGCT ADCYAP1R1 MIR550B1 LINC01176 CCDC129 GARS AQP1 LSM5 FAM188B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |