Assay Details
Target Gene Details
Entrez Gene ID: | 223075 |
Gene Name: | coiled-coil domain containing 129 |
Gene Aliases: |
- |
Location: |
Chr.7:31514071-31658720 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CCDC129 | NM_001257967.1 | NP_001244896.1 | ||
NM_001257968.1 | NP_001244897.1 | |||
NR_047565.1 | ||||
XM_011515213.1 | XP_011513515.1 | |||
XM_017011871.1 | XP_016867360.1 | |||
XM_017011872.1 | XP_016867361.1 | |||
AK128026.1 | BAC87235.1 | |||
AK295011.1 | ||||
AK304387.1 | ||||
BC132719.1 | ||||
DA098735.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3891094 | Chr.7:31500343 - 31591980 on Build GRCh38 | Gain | CCDC129 |
nsv436557 | Chr.7:30288062 - 32730902 on Build GRCh38 | Deletion | MIR550B1 GHRHR ZNRF2 PDE1C MIR550A1 PPP1R17 INMT ADCYAP1R1 FAM188B DKFZP586I1420 CRHR2 DPY19L1P1 LOC100130673 GARS AVL9 CCDC129 AQP1 ZNRF2P1 LINC01176 NOD1 GGCT LOC401320 INMT-FAM188B NEUROD6 LSM5 LOC101928268 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |