Assay Details
Target Gene Details
Entrez Gene ID: | 100505881 |
Gene Name: | MAGI2 antisense RNA 3 |
Gene Aliases: |
- |
Location: |
Chr.7:79452957-79471208 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAGI2-AS3 | NR_038343.2 | |||
NR_038344.1 | ||||
NR_038345.1 | ||||
NR_038346.1 | ||||
AB371456.1 | ||||
AK311103.1 | ||||
BI521274.1 | ||||
BI868593.1 | ||||
CN311982.1 | ||||
CR749529.1 | ||||
DA758742.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv607659 | Chr.7:78863326 - 79854164 on Build GRCh38 | Gain | MAGI2-AS3 MAGI2-AS2 MAGI2 |
esv2764028 | Chr.7:79245424 - 79483314 on Build GRCh38 | Gain+Loss | MAGI2-AS3 MAGI2 |