Assay Details
Target Gene Details
Entrez Gene ID: | 10652 |
Gene Name: | YKT6 v-SNARE homolog (S. cerevisiae) |
Gene Aliases: |
- |
Location: |
Chr.7:44200968-44214294 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
YKT6 | NM_006555.3 | NP_006546.1 | ||
XM_005249582.4 | XP_005249639.1 | |||
XM_017011682.1 | XP_016867171.1 | |||
AB451317.1 | ||||
AB451447.1 | ||||
AF091079.1 | AAC72948.1 | |||
AK223488.1 | BAD97208.1 | |||
AK299158.1 | ||||
AK312622.1 | ||||
AU130339.1 | ||||
BC007319.2 | AAH07319.1 | |||
BT007078.1 | AAP35741.1 | |||
CR456989.1 | CAG33270.1 | |||
CR542008.1 | CAG46805.1 | |||
U95735.1 | AAB81131.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv8082 | Chr.7:43956156 - 44207229 on Build GRCh38 | Gain | DBNL POLR2J4 AEBP1 POLD2 PGAM2 MIR4649 MIR6837 POLM YKT6 RASA4CP MYL7 MIR6838 LINC00957 SPDYE1 GCK |
nsv509195 | Chr.7:44147118 - 44261782 on Build GRCh38 | Insertion | CAMK2B YKT6 GCK |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |