Assay Details
Target Gene Details
Entrez Gene ID: | 10926 |
Gene Name: | DBF4 zinc finger |
Gene Aliases: |
ASK, CHIF, DBF4A, ZDBF1 |
Location: |
Chr.7:87876229-87909541 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DBF4 | NM_001318060.1 | NP_001304989.1 | ||
NM_001318061.1 | NP_001304990.1 | |||
NM_001318062.1 | NP_001304991.1 | |||
NM_006716.3 | NP_006707.1 | |||
AB028069.1 | BAA78326.1 | |||
AB028070.1 | BAA78327.1 | |||
AF160249.1 | AAD41911.1 | |||
AF160876.1 | AAD45357.1 | |||
AK292569.1 | ||||
AK302010.1 | ||||
AK303175.1 | ||||
BC036045.1 | AAH36045.1 | |||
BC047693.1 | AAH47693.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464622 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain | TMEM243 SLC25A40 CROT DBF4 ABCB4 TP53TG1 RUNDC3B ABCB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |