Assay Details
Target Gene Details
Entrez Gene ID: | 1021 |
Gene Name: | cyclin dependent kinase 6 |
Gene Aliases: |
MCPH12, PLSTIRE |
Location: |
Chr.7:92604921-92836627 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CDK6 | NM_001145306.1 | NP_001138778.1 | ||
NM_001259.6 | NP_001250.1 | |||
XM_006715835.2 | XP_006715898.1 | |||
AK313491.1 | ||||
BC052264.1 | AAH52264.1 | |||
X66365.1 | CAA47008.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv831062 | Chr.7:92611920 - 92831297 on Build GRCh38 | Gain | CDK6 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |