Assay Details
Target Gene Details
Entrez Gene ID: | 54677 |
Gene Name: | carnitine O-octanoyltransferase |
Gene Aliases: |
COT |
Location: |
Chr.7:87345635-87399796 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CROT | NM_001143935.1 | NP_001137407.1 | ||
NM_021151.3 | NP_066974.2 | |||
XM_011516337.2 | XP_011514639.1 | |||
XM_017012370.1 | XP_016867859.1 | |||
AF073770.1 | AAD41654.1 | |||
AF168793.1 | AAF03234.1 | |||
AK127901.1 | ||||
AK300114.1 | ||||
AK313487.1 | ||||
BC039004.1 | AAH39004.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv464622 | Chr.7:87203287 - 87911933 on Build GRCh38 | Gain | DBF4 ABCB4 TMEM243 SLC25A40 RUNDC3B ABCB1 TP53TG1 CROT |
nsv428177 | Chr.7:87228237 - 87404851 on Build GRCh38 | Loss | ABCB4 TP53TG1 CROT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |