Assay Details
Target Gene Details
Entrez Gene ID: | 57157 |
Gene Name: | putative homeodomain transcription factor 2 |
Gene Aliases: |
- |
Location: |
Chr.7:77798792-77957504 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PHTF2 | NM_001127358.1 | NP_001120830.1 | ||
NM_001127360.1 | NP_001120832.1 | |||
XM_005250508.4 | XP_005250565.1 | |||
XM_005250509.4 | XP_005250566.1 | |||
XM_006716063.3 | XP_006716126.1 | |||
XM_011516422.2 | XP_011514724.1 | |||
XM_011516423.2 | XP_011514725.1 | |||
XM_017012458.1 | XP_016867947.1 | |||
XM_017012459.1 | XP_016867948.1 | |||
AK090643.1 | ||||
AK225854.1 | ||||
AK308289.1 | ||||
AL136883.1 | CAB66817.1 | |||
BC067742.1 | AAH67742.1 | |||
BP321440.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)