Assay Details
Target Gene Details
Entrez Gene ID: | 29116 |
Gene Name: | myosin regulatory light chain interacting protein |
Gene Aliases: |
IDOL, MIR |
Location: |
Chr.6:16129086-16151015 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MYLIP | NM_013262.3 | NP_037394.2 | ||
XM_005249032.2 | XP_005249089.1 | |||
XM_005249033.2 | XP_005249090.1 | |||
XM_017010789.1 | XP_016866278.1 | |||
AF006003.1 | AAQ13408.1 | |||
AF187016.2 | AAF18974.1 | |||
AF212221.1 | AAF87323.1 | |||
AF258586.1 | AAG23789.1 | |||
AK074391.1 | ||||
AK314648.1 | ||||
BC002860.2 | AAH02860.1 | |||
BT007055.1 | AAP35704.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv950132 | Chr.6:16128770 - 16148769 on Build GRCh38 | Duplication | MYLIP MIR4639 |
nsv5209 | Chr.6:16117893 - 16151799 on Build GRCh38 | Insertion | MYLIP MIR4639 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |