Assay Details
Target Gene Details
Entrez Gene ID: | 2762 |
Gene Name: | GDP-mannose 4,6-dehydratase |
Gene Aliases: |
GMD, SDR3E1 |
Location: |
Chr.6:1623800-2245634 on Build GRCh38 |
Assay Gene Location: | Within Intron 17 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GMDS | NM_001253846.1 | NP_001240775.1 | ||
NM_001500.3 | NP_001491.1 | |||
XM_011514500.1 | XP_011512802.1 | |||
XM_017010752.1 | XP_016866241.1 | |||
AF040260.1 | AAC24501.1 | |||
AF042377.1 | AAC13553.1 | |||
AI761361.1 | ||||
AK057484.1 | ||||
AK313953.1 | ||||
BC000117.1 | AAH00117.1 | |||
CR541929.1 | CAG46727.1 | |||
CR541947.1 | CAG46745.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv992n67 | Chr.6:1589856 - 1629286 on Build GRCh38 | Gain | FOXC1 GMDS FOXCUT |
nsv1016859 | Chr.6:1459785 - 1645585 on Build GRCh38 | Gain | FOXC1 GMDS FOXCUT |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |