Assay Details
Target Gene Details
Entrez Gene ID: | 3354 |
Gene Name: | 5-hydroxytryptamine receptor 1E |
Gene Aliases: |
5-HT1E |
Location: |
Chr.6:86936919-87020068 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HTR1E | NM_000865.2 | NP_000856.1 | ||
XM_011535789.2 | XP_011534091.1 | |||
AL521170.3 | ||||
BC069751.1 | AAH69751.1 | |||
M91467.1 | AAA58353.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv604106 | Chr.6:85995537 - 87138584 on Build GRCh38 | Gain | HTR1E LOC107986532 CGA |
esv2763585 | Chr.6:85938269 - 87211344 on Build GRCh38 | Gain | ZNF292 HTR1E LOC107986532 CGA |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |