Assay Details
Target Gene Details
Entrez Gene ID: | 9532 |
Gene Name: | BCL2 associated athanogene 2 |
Gene Aliases: |
BAG-2, dJ417I1.2 |
Location: |
Chr.6:57172306-57185215 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
BAG2 | NM_004282.3 | NP_004273.1 | ||
XM_005249490.3 | XP_005249547.1 | |||
XM_011514998.2 | XP_011513300.1 | |||
XM_011514999.2 | XP_011513301.1 | |||
AF095192.1 | AAD16121.1 | |||
AK001015.1 | ||||
AK023735.1 | ||||
AK301934.1 | ||||
AL050287.1 | CAB43388.1 | |||
BC125039.1 | ||||
CR533496.1 | CAG38527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv462949 | Chr.6:57109104 - 57234175 on Build GRCh38 | Loss | LOC101927211 BAG2 RAB23 ZNF451 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |