Assay Details
Target Gene Details
Entrez Gene ID: | 6885 |
Gene Name: | mitogen-activated protein kinase kinase kinase 7 |
Gene Aliases: |
MEKK7, TAK1, TGF1a |
Location: |
Chr.6:90513573-90587301 on Build GRCh38 |
Assay Gene Location: | Within Intron 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MAP3K7 | NM_003188.3 | NP_003179.1 | ||
NM_145331.2 | NP_663304.1 | |||
NM_145332.2 | NP_663305.1 | |||
NM_145333.2 | NP_663306.1 | |||
XM_006715553.2 | XP_006715616.1 | |||
XM_017011226.1 | XP_016866715.1 | |||
AB009356.1 | BAA25025.1 | |||
AB009357.1 | BAA25026.1 | |||
AB009358.2 | BAA25027.2 | |||
AF218074.1 | AAF27652.1 | |||
AK055901.1 | ||||
AK299268.1 | ||||
AK315774.1 | ||||
AL050393.1 | CAB43687.2 | |||
BC017715.2 | AAH17715.1 | |||
BT019654.1 | AAV38460.1 | |||
BT019655.1 | AAV38461.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv523939 | Chr.6:90357904 - 91080775 on Build GRCh38 | Gain |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)