Assay Details
Target Gene Details
Entrez Gene ID: | 27242 |
Gene Name: | TNF receptor superfamily member 21 |
Gene Aliases: |
BM-018, CD358, DR6 |
Location: |
Chr.6:47231527-47309976 on Build GRCh38 |
Assay Gene Location: | Within Intron 5 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TNFRSF21 | NM_014452.4 | NP_055267.1 | ||
AB209394.1 | BAD92631.1 | |||
AF068868.1 | AAC34583.1 | |||
AJ420531.1 | ||||
AK001504.1 | ||||
AK315560.1 | ||||
AY358304.1 | AAQ88671.1 | |||
BC010241.1 | AAH10241.1 | |||
BC017730.2 | AAH17730.1 | |||
BC021572.1 | ||||
BT007420.1 | AAP36088.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1017113 | Chr.6:46846140 - 48020279 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)