Assay Details
Target Gene Details
Entrez Gene ID: | 1909 |
Gene Name: | endothelin receptor type A |
Gene Aliases: |
ET-A, ETA, ETA-R, ETAR, ETRA, MFDA, hET-AR |
Location: |
Chr.4:147480917-147544954 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EDNRA | NM_001166055.1 | NP_001159527.1 | ||
NM_001256283.1 | NP_001243212.1 | |||
NM_001957.3 | NP_001948.1 | |||
NR_045958.1 | ||||
AF014826.1 | AAB94859.1 | |||
AK304451.1 | ||||
AK312812.1 | ||||
AK315931.1 | ||||
AY275462.1 | AAP32294.1 | |||
BC022511.1 | AAH22511.1 | |||
D90348.1 | BAA14359.1 | |||
L06622.1 | AAA58447.1 | |||
S45956.1 | AAB23644.1 | |||
S57498.1 | AAB25530.2 | |||
S63938.1 | AAB20278.1 | |||
S67127.1 | AAB20407.1 | |||
S81539.1 | AAB36325.1 | |||
S81542.1 | AAB36326.1 | |||
S81545.1 | AAB36327.1 | |||
X61950.1 | CAA43953.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830109 | Chr.4:147373909 - 147547453 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)