Assay Details
Target Gene Details
Entrez Gene ID: | 3490 |
Gene Name: | insulin like growth factor binding protein 7 |
Gene Aliases: |
AGM, FSTL2, IBP-7, IGFBP-7, IGFBP-7v, IGFBPRP1, MAC25, PSF, RAMSVPS, TAF |
Location: |
Chr.4:57031071-57110385 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IGFBP7 | NM_001253835.1 | NP_001240764.1 | ||
NM_001553.2 | NP_001544.1 | |||
AF540057.1 | AAN23122.1 | |||
AK303915.1 | ||||
AK316082.1 | ||||
BC017201.2 | AAH17201.1 | |||
BC066339.1 | AAH66339.1 | |||
BT006654.1 | AAP35300.1 | |||
L19182.1 | AAA16187.1 | |||
S75725.1 | AAB32370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv1483e212 | Chr.4:57035151 - 57057045 on Build GRCh38 | Loss |
![]() |
esv3569435 | Chr.4:57040752 - 57057045 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)