Assay Details
Target Gene Details
Entrez Gene ID: | 101928809 |
Gene Name: | uncharacterized LOC101928809 |
Gene Aliases: |
- |
Location: |
Chr.4:75980790-76005942 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LOC101928809 | NR_125906.1 |