Assay Details
Target Gene Details
Entrez Gene ID: | 9255 |
Gene Name: | aminoacyl tRNA synthetase complex interacting multifunctional protein 1 |
Gene Aliases: |
EMAP2, EMAPII, HLD3, SCYE1, p43 |
Location: |
Chr.4:106315610-106349225 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
AIMP1 | NM_001142415.1 | NP_001135887.1 | ||
NM_001142416.1 | NP_001135888.1 | |||
NM_004757.3 | NP_004748.2 | |||
XM_017008835.1 | XP_016864324.1 | |||
XM_017008836.1 | XP_016864325.1 | |||
AK095951.1 | ||||
AK297950.1 | ||||
AK303965.1 | ||||
BC014051.2 | AAH14051.1 | |||
BI258927.1 | ||||
CR542281.1 | CAG47076.1 | |||
U10117.1 | AAA62202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830029 | Chr.4:106281939 - 106432326 on Build GRCh38 | Gain | TBCK AIMP1 GIMD1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |