Assay Details
Target Gene Details
Entrez Gene ID: | 79658 |
Gene Name: | Rho GTPase activating protein 10 |
Gene Aliases: |
GRAF2, PS-GAP, PSGAP |
Location: |
Chr.4:147732063-148072781 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ARHGAP10 | NM_024605.3 | NP_078881.3 | ||
XM_005263215.3 | XP_005263272.1 | |||
XM_017008602.1 | XP_016864091.1 | |||
AB050785.1 | BAB61771.1 | |||
AK123785.1 | ||||
BC109029.2 | AAI09030.1 | |||
BC109030.1 | AAI09031.1 | |||
BC126899.1 | ||||
BC128055.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3602608 | Chr.4:147742992 - 147755464 on Build GRCh38 | Loss |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)