Assay Details
Target Gene Details
Entrez Gene ID: | 92597 |
Gene Name: | MOB kinase activator 1B |
Gene Aliases: |
MATS2, MOB4A, MOBKL1A |
Location: |
Chr.4:70901970-70988174 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MOB1B | NM_001244766.1 | NP_001231695.1 | ||
NM_001244767.1 | NP_001231696.1 | |||
NM_173468.3 | NP_775739.1 | |||
XM_005265709.2 | XP_005265766.1 | |||
XM_011532412.2 | XP_011530714.1 | |||
XM_017008837.1 | XP_016864326.1 | |||
AK093545.1 | ||||
AK299481.1 | ||||
AK313513.1 | ||||
BC038112.1 | AAH38112.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain | UTP3 FDCSP ODAM CSN3 MUC7 HTN3 SMR3A AMBN CABS1 ENAM MOB1B HTN1 RUFY3 SMR3B CSN1S2AP CSN1S2BP OPRPN JCHAIN GRSF1 AMTN PRR27 STATH |
nsv829966 | Chr.4:70794825 - 70949164 on Build GRCh38 | Loss | RUFY3 GRSF1 MOB1B |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |