Assay Details
Target Gene Details
Entrez Gene ID: | 3660 |
Gene Name: | interferon regulatory factor 2 |
Gene Aliases: |
IRF-2 |
Location: |
Chr.4:184387722-184474572 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
IRF2 | NM_002199.3 | NP_002190.2 | ||
AK312953.1 | ||||
BC015803.2 | AAH15803.1 | |||
BT007264.1 | AAP35928.1 | |||
CR457077.1 | CAG33358.1 | |||
X15949.1 | CAA34073.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1026074 | Chr.4:184384963 - 184780663 on Build GRCh38 | Gain | CENPU ACSL1 PRIMPOL IRF2 LVCAT8 CASP3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |