Assay Details
Target Gene Details
Entrez Gene ID: | 2868 |
Gene Name: | G protein-coupled receptor kinase 4 |
Gene Aliases: |
GPRK2L, GPRK4, GRK4a, IT11 |
Location: |
Chr.4:2963504-3040760 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3575673 | Chr.4:2957494 - 3022996 on Build GRCh38 | Gain |
![]() ![]() |
dgv5082n100 | Chr.4:2956087 - 3017315 on Build GRCh38 | Gain |
![]() ![]() |
nsv593438 | Chr.4:2946729 - 3028370 on Build GRCh38 | Gain |
![]() ![]() ![]() |
esv3599415 | Chr.4:2958013 - 3019896 on Build GRCh38 | Gain |
![]() ![]() |
esv3599412 | Chr.4:2940424 - 3030117 on Build GRCh38 | Gain |
![]() ![]() ![]() |
nsv593437 | Chr.4:2904558 - 3233253 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)