Assay Details
Target Gene Details
Entrez Gene ID: | 57620 |
Gene Name: | stromal interaction molecule 2 |
Gene Aliases: |
- |
Location: |
Chr.4:26860691-27025381 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STIM2 | NM_001169117.1 | NP_001162588.1 | ||
NM_001169118.1 | NP_001162589.1 | |||
NM_020860.3 | NP_065911.3 | |||
AB040915.1 | ||||
AF328905.1 | AAK82337.1 | |||
AK096846.1 | ||||
AL136577.1 | CAB66512.2 | |||
BC136449.1 | ||||
BC146661.1 | ||||
BC152554.1 | ||||
BC171766.1 | ||||
BX343313.2 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593872 | Chr.4:26843052 - 27076754 on Build GRCh38 | Loss | LOC105374546 STIM2 TBC1D19 |
nsv470021 | Chr.4:26719956 - 27065405 on Build GRCh38 | Loss | LOC105374546 STIM2 TBC1D19 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |