Assay Details
Target Gene Details
Entrez Gene ID: | 10891 |
Gene Name: | PPARG coactivator 1 alpha |
Gene Aliases: |
LEM6, PGC-1(alpha), PGC-1alpha, PGC-1v, PGC1, PGC1A, PPARGC1 |
Location: |
Chr.4:23792021-24472975 on Build GRCh38 |
Assay Gene Location: | Within Intron 22 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PPARGC1A | NM_013261.3 | NP_037393.1 | ||
XM_005248131.4 | XP_005248188.1 | |||
XM_005248132.1 | XP_005248189.1 | |||
XM_005248134.4 | XP_005248191.1 | |||
XM_011513765.2 | XP_011512067.1 | |||
XM_011513766.1 | XP_011512068.1 | |||
XM_011513767.2 | XP_011512069.1 | |||
XM_011513768.1 | XP_011512070.1 | |||
XM_011513770.2 | XP_011512072.1 | |||
XM_011513771.1 | XP_011512073.1 | |||
XM_017007664.1 | XP_016863153.1 | |||
AB061325.1 | BAE46508.1 | |||
AF106698.1 | AAF18573.1 | |||
AF159714.1 | AAD51615.1 | |||
AF186379.1 | AAD56250.1 | |||
HQ695733.1 | ||||
JQ772116.1 | ||||
JQ772117.1 | ||||
JQ772118.1 | ||||
JQ772119.1 | ||||
JQ772120.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv593825 | Chr.4:23700935 - 23888959 on Build GRCh38 | Gain | PPARGC1A |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |