Assay Details
Target Gene Details
Entrez Gene ID: | 7350 |
Gene Name: | uncoupling protein 1 |
Gene Aliases: |
SLC25A7, UCP |
Location: |
Chr.4:140555770-140568914 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
UCP1 | NM_021833.4 | NP_068605.1 | ||
XM_005263206.3 | XP_005263263.1 | |||
XM_011532228.2 | XP_011530530.1 | |||
BC069556.1 | AAH69556.1 | |||
BC096736.1 | AAH96736.1 | |||
BC098168.1 | AAH98168.1 | |||
BC098258.1 | AAH98258.1 | |||
BC098352.1 | AAH98352.1 | |||
U28480.1 | AAA85271.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830088 | Chr.4:140436956 - 140603794 on Build GRCh38 | Gain | MGAT4D ELMOD2 UCP1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |