Assay Details
Target Gene Details
Entrez Gene ID: | 353497 |
Gene Name: | DNA polymerase nu |
Gene Aliases: |
POL4P |
Location: |
Chr.4:2071958-2242164 on Build GRCh38 |
Assay Gene Location: | Within Intron 21 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLN | NM_181808.3 | NP_861524.2 | ||
AF044578.1 | AAD02338.1 | |||
AK131239.1 | BAD18421.1 | |||
AY136549.1 | AAN52116.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)