Assay Details
Target Gene Details
Entrez Gene ID: | 101927186 |
Gene Name: | adhesion G protein-coupled receptor L3 antisense RNA 1 |
Gene Aliases: |
LPHN3-AS1 |
Location: |
Chr.4:62071755-62161765 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ADGRL3-AS1 | NR_110595.1 | |||
DB518756.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1012574 | Chr.4:61978546 - 62088469 on Build GRCh38 | Gain |
![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)