Assay Details
Target Gene Details
Entrez Gene ID: | 10463 |
Gene Name: | solute carrier family 30 member 9 |
Gene Aliases: |
C4orf1, GAC63, HUEL, ZNT9 |
Location: |
Chr.4:41990472-42087534 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SLC30A9 | NM_006345.3 | NP_006336.3 | ||
XM_017007654.1 | XP_016863143.1 | |||
AF006621.2 | AAB87763.2 | |||
AK225470.1 | ||||
AK299917.1 | ||||
AK312838.1 | ||||
AY319413.1 | AAP83846.1 | |||
AY594282.1 | AAT02479.1 | |||
BC007732.2 | AAH07732.1 | |||
BC016949.1 | AAH16949.1 | |||
KU178526.1 | ||||
KU178527.1 |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)