Assay Details
Target Gene Details
Entrez Gene ID: | 79441 |
Gene Name: | HAUS augmin like complex subunit 3 |
Gene Aliases: |
C4orf15, IT1, dgt3 |
Location: |
Chr.4:2231836-2242164 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
HAUS3 | NM_001303143.1 | NP_001290072.1 | ||
NM_024511.6 | NP_078787.2 | |||
AF040964.1 | AAB97010.1 | |||
AK056134.1 | ||||
AK293948.1 | ||||
BC003648.2 | AAH03648.1 | |||
BC025356.1 | AAH25356.1 | |||
BF508642.1 | ||||
CR749640.1 | CAH18434.1 |
Target Gene Details
Entrez Gene ID: | 353497 |
Gene Name: | DNA polymerase nu |
Gene Aliases: |
POL4P |
Location: |
Chr.4:2071958-2242164 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
POLN | NM_181808.3 | NP_861524.2 | ||
AK303673.1 | ||||
DB098342.1 | ||||
HY010559.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1009347 | Chr.4:2200935 - 2516063 on Build GRCh38 | Gain | ZFYVE28 RNF4 MIR4800 MXD4 CFAP99 POLN HAUS3 |
esv2726835 | Chr.4:2019561 - 2959568 on Build GRCh38 | Deletion | NOP14 ZFYVE28 RNF4 MIR4800 SH3BP2 C4orf48 FAM193A CFAP99 ADD1 TNIP2 NOP14-AS1 NAT8L MFSD10 MXD4 POLN HAUS3 |
nsv829838 | Chr.4:2155167 - 2349950 on Build GRCh38 | Loss | ZFYVE28 MIR4800 MXD4 POLN HAUS3 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |