Assay Details
Target Gene Details
Entrez Gene ID: | 3512 |
Gene Name: | joining chain of multimeric IgA and IgM |
Gene Aliases: |
IGCJ, IGJ, JCH |
Location: |
Chr.4:70655541-70676901 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
JCHAIN | NM_144646.3 | NP_653247.1 | ||
XM_006714211.2 | XP_006714274.1 | |||
XM_006714212.2 | XP_006714275.1 | |||
XM_011531925.1 | XP_011530227.1 | |||
XM_011531926.1 | XP_011530228.1 | |||
AK312014.1 | ||||
BC038982.1 | AAH38982.1 | |||
BP261870.1 | ||||
DQ884395.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv829965 | Chr.4:70630001 - 70806591 on Build GRCh38 | Gain | JCHAIN ENAM UTP3 RUFY3 |
nsv1012495 | Chr.4:69979953 - 70947088 on Build GRCh38 | Gain | MOB1B ODAM SMR3B CSN3 CSN1S2AP AMBN OPRPN PRR27 HTN3 CABS1 GRSF1 MUC7 AMTN JCHAIN SMR3A HTN1 CSN1S2BP FDCSP ENAM UTP3 RUFY3 STATH |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |