Assay Details
Target Gene Details
Entrez Gene ID: | 55351 |
Gene Name: | serine/threonine kinase 32B |
Gene Aliases: |
HSA250839, STK32, STKG6, YANK2 |
Location: |
Chr.4:5051323-5501001 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
STK32B | NM_001306082.1 | NP_001293011.1 | ||
NM_018401.2 | NP_060871.1 | |||
XM_011513497.1 | XP_011511799.1 | |||
AJ250839.1 | CAB76471.1 | |||
AK292696.1 | ||||
AK313846.1 | ||||
BC038238.1 | AAH38238.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1011885 | Chr.4:5091796 - 5119723 on Build GRCh38 | Loss |
![]() |
nsv593479 | Chr.4:4899358 - 5459331 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() |
nsv997889 | Chr.4:4785891 - 5220458 on Build GRCh38 | Gain |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
nsv593480 | Chr.4:4961200 - 5760434 on Build GRCh38 | Loss |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)