Assay Details
Target Gene Details
Entrez Gene ID: | 8001 |
Gene Name: | glycine receptor alpha 3 |
Gene Aliases: |
- |
Location: |
Chr.4:174642011-174829568 on Build GRCh38 |
Assay Gene Location: | Within Intron 12 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GLRA3 | NM_001042543.2 | NP_001036008.1 | ||
NM_006529.3 | NP_006520.2 | |||
XM_011532268.2 | XP_011530570.1 | |||
XM_017008630.1 | XP_016864119.1 | |||
AF018157.1 | AAC39918.1 | |||
BC036086.1 | AAH36086.1 | |||
U93917.1 | AAC39917.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |