Assay Details
Target Gene Details
Entrez Gene ID: | 9575 |
Gene Name: | clock circadian regulator |
Gene Aliases: |
KAT13D, bHLHe8 |
Location: |
Chr.4:55427901-55547138 on Build GRCh38 |
Assay Gene Location: | Within Intron 27 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CLOCK | NM_001267843.1 | NP_001254772.1 | ||
NM_004898.3 | NP_004889.1 | |||
XM_005265787.2 | XP_005265844.1 | |||
XM_011534409.2 | XP_011532711.1 | |||
XM_011534410.2 | XP_011532712.1 | |||
XM_011534411.2 | XP_011532713.1 | |||
XM_017008854.1 | XP_016864343.1 | |||
XM_017008855.1 | XP_016864344.1 | |||
AB002332.1 | ||||
AF011568.1 | AAB83969.1 | |||
AK124437.1 | 1 | 2736 | ||
AK223549.1 | BAD97269.1 | |||
AK291708.1 | ||||
BC041878.1 | AAH41878.1 | |||
BC126157.1 | ||||
BC126159.1 | ||||
BX537616.1 |
Target Gene Details
Entrez Gene ID: | 55858 |
Gene Name: | transmembrane protein 165 |
Gene Aliases: |
CDG2K, FT27, GDT1, TMPT27, TPARL |
Location: |
Chr.4:55395913-55453397 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TMEM165 | XM_011534394.2 | XP_011532696.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |