Assay Details
Target Gene Details
Entrez Gene ID: | 152586 |
Gene Name: | MGAT4 family member D |
Gene Aliases: |
GnT1IP |
Location: |
Chr.4:140442257-140499829 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MGAT4D | NM_001277353.1 | NP_001264282.1 | ||
XM_011531650.2 | XP_011529952.1 | |||
XM_011531651.2 | XP_011529953.1 | |||
XM_011531652.2 | XP_011529954.1 | |||
XM_011531653.2 | XP_011529955.1 | |||
XM_011531654.2 | XP_011529956.1 | |||
XM_011531656.2 | XP_011529958.1 | |||
XM_011531657.2 | XP_011529959.1 | |||
XM_011531658.2 | XP_011529960.1 | |||
XM_011531659.1 | XP_011529961.1 | |||
XM_011531661.1 | XP_011529963.1 | |||
XM_011531662.1 | XP_011529964.1 | |||
XM_011531663.2 | XP_011529965.1 | |||
XM_011531664.2 | XP_011529966.1 | |||
XM_011531665.2 | XP_011529967.1 | |||
BI826002.1 | ||||
BI828214.1 | ||||
DB444046.1 | ||||
DB462368.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv830088 | Chr.4:140436956 - 140603794 on Build GRCh38 | Gain | UCP1 MGAT4D ELMOD2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |