Assay Details
Target Gene Details
Entrez Gene ID: | 641365 |
Gene Name: | long intergenic non-protein coding RNA 616 |
Gene Aliases: |
- |
Location: |
Chr.4:138027422-138130685 on Build GRCh38 |
Assay Gene Location: | Within Intron 9 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LINC00616 | NR_037866.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1033022 | Chr.4:137999933 - 138081308 on Build GRCh38 | Loss | LINC00616 |