Assay Details
Target Gene Details
Entrez Gene ID: | 81579 |
Gene Name: | phospholipase A2 group XIIA |
Gene Aliases: |
GXII, PLA2G12, ROSSY |
Location: |
Chr.4:109709989-109730086 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
PLA2G12A | NM_030821.4 | NP_110448.2 | ||
AF306567.1 | AAG50243.1 | |||
AF332892.1 | AAG50289.1 | |||
AK075389.1 | BAC11589.1 | |||
AK313081.1 | ||||
AY359024.1 | AAQ89383.1 | |||
BC017218.2 | AAH17218.1 | |||
BI667755.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv595142 | Chr.4:109688325 - 109735574 on Build GRCh38 | Loss | CASP6 PLA2G12A CFI MCUB |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |