Assay Details
Target Gene Details
Entrez Gene ID: | 54532 |
Gene Name: | ubiquitin specific peptidase 53 |
Gene Aliases: |
- |
Location: |
Chr.4:119212583-119295518 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
USP53 | NM_019050.2 | NP_061923.2 | ||
XM_005263073.3 | XP_005263130.1 | |||
XM_005263075.2 | XP_005263132.1 | |||
XM_005263076.2 | XP_005263133.1 | |||
XM_005263077.3 | XP_005263134.1 | |||
XM_006714240.1 | XP_006714303.1 | |||
XM_011532037.2 | XP_011530339.1 | |||
XM_011532038.2 | XP_011530340.1 | |||
XM_017008312.1 | XP_016863801.1 | |||
XM_017008313.1 | XP_016863802.1 | |||
XM_017008314.1 | XP_016863803.1 | |||
XM_017008315.1 | XP_016863804.1 | |||
XM_017008316.1 | XP_016863805.1 | |||
XM_017008317.1 | XP_016863806.1 | |||
XM_017008318.1 | XP_016863807.1 | |||
BX648836.1 | ||||
CR749490.1 | CAH18315.1 | |||
DA859085.1 | ||||
DA886370.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv5373n100 | Chr.4:119086758 - 119222694 on Build GRCh38 | Gain | USP53 LOC101929762 MYOZ2 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |