Assay Details
Target Gene Details
Entrez Gene ID: | 55601 |
Gene Name: | DEXD/H-box helicase 60 |
Gene Aliases: |
- |
Location: |
Chr.4:168216291-168318807 on Build GRCh38 |
Assay Gene Location: | Within Intron 31 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
DDX60 | NM_017631.5 | NP_060101.3 | ||
XM_011532103.2 | XP_011530405.1 | |||
AK000042.1 | BAA90901.1 | |||
AK223184.1 | BAD96904.1 | |||
AK225191.1 | ||||
BC038115.2 | AAH38115.1 | |||
BC074781.2 | AAH74781.1 | |||
BX648655.1 | CAE46197.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4596 | Chr.4:168228336 - 168263735 on Build GRCh38 | Insertion | DDX60 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |