Assay Details
Target Gene Details
Entrez Gene ID: | 2047 |
Gene Name: | EPH receptor B1 |
Gene Aliases: |
ELK, EPHT2, Hek6, NET |
Location: |
Chr.3:134776089-135260467 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
EPHB1 | NM_004441.4 | NP_004432.1 | ||
XM_017005866.1 | XP_016861355.1 | |||
XM_017005868.1 | XP_016861357.1 | |||
AF037333.1 | AAB94627.1 | |||
AF037334.1 | AAB94628.1 | |||
AK291208.1 | ||||
AK309741.1 | ||||
BC111744.1 | ||||
DA107464.1 | ||||
EU826607.1 | ||||
L40636.1 | AAB08520.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv508956 | Chr.3:134808988 - 134822762 on Build GRCh38 | Insertion | EPHB1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |