Assay Details
Target Gene Details
Entrez Gene ID: | 2960 |
Gene Name: | general transcription factor IIE subunit 1 |
Gene Aliases: |
FE, TF2E1, TFIIE-A |
Location: |
Chr.3:120742711-120783134 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GTF2E1 | NM_005513.2 | NP_005504.2 | ||
XM_011512745.2 | XP_011511047.1 | |||
AK223401.1 | BAD97121.1 | |||
BC005917.1 | ||||
DA498425.1 | ||||
S67859.1 | AAB20413.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
dgv4847n100 | Chr.3:120741894 - 120864319 on Build GRCh38 | Gain | RABL3 LOC101926983 GTF2E1 |
nsv829699 | Chr.3:120589485 - 120765848 on Build GRCh38 | Loss | RABL3 NDUFB4 HGD GTF2E1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |