Assay Details
Target Gene Details
Entrez Gene ID: | 152189 |
Gene Name: | CKLF like MARVEL transmembrane domain containing 8 |
Gene Aliases: |
CKLFSF8, CKLFSF8-V2 |
Location: |
Chr.3:32238679-32370325 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
CMTM8 | NM_001320308.1 | NP_001307237.1 | ||
NM_178868.4 | NP_849199.2 | |||
XM_011533416.2 | XP_011531718.1 | |||
AF474370.2 | AAN73034.1 | |||
AY598783.1 | ||||
BC041390.2 | AAH41390.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv1003285 | Chr.3:31386933 - 33172886 on Build GRCh38 | Gain | TRIM71 CMTM7 CRTAP CCR4 ZNF860 DYNC1LI1 SUSD5 CMTM6 TMPPE CMTM8 OSBPL10 CNOT10 MIR548AY LOC105377021 GPD1L GLB1 LOC105377022 STT3B OSBPL10-AS1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic DGV Variation |