Assay Details
Target Gene Details
Entrez Gene ID: | 8626 |
Gene Name: | tumor protein p63 |
Gene Aliases: |
AIS, B(p51A), B(p51B), EEC3, KET, LMS, NBP, OFC8, RHS, SHFM4, TP53CP, TP53L, TP73L, p40, p51, p53CP, p63, p73H, p73L |
Location: |
Chr.3:189597115-189897279 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
TP63 | XM_011513251.2 | XP_011511553.1 | ||
XM_011513252.2 | XP_011511554.1 |
More Information
Set Membership: |
Intragenic Intronic Non-exonic non-DGV Variation |