Assay Details
Target Gene Details
Entrez Gene ID: | 59343 |
Gene Name: | SUMO1/sentrin/SMT3 specific peptidase 2 |
Gene Aliases: |
AXAM2, SMT3IP2 |
Location: |
Chr.3:185586228-185631101 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
SENP2 | NM_021627.2 | NP_067640.2 | ||
XM_005247690.2 | XP_005247747.2 | |||
XM_005247691.2 | XP_005247748.1 | |||
XM_017006973.1 | XP_016862462.1 | |||
AF151697.2 | AAG15309.2 | |||
AK027599.1 | BAB55222.1 | |||
AK296744.1 | ||||
AK296772.1 | ||||
AK298628.1 | ||||
AK304402.1 | ||||
AL834380.1 | CAD39043.1 | |||
BC040609.1 | AAH40609.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
nsv4148 | Chr.3:185592906 - 185625116 on Build GRCh38 | Insertion |
![]() |
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)
More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
![](/order/genome-database/details/genotyping/assets/img/arrow_icon_up_black.png)